Learn More
Macherey-Nagel Bioanalysis™ NucleoSpin™ 96 Gel and PCR Clean-Up Kit
NucleoSpin™ 96 Gel and PCR Clean-Up Kit provides time saving medium to high throughput PCR clean up.
239.00 € - 3040.00 €
Specifications
Content And Storage | 18°C to 25°C |
---|---|
Format | 96-well Plate |
For Use With (Application) | PCR clean up |
Sample Volume | < 100 μL |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
12718512
|
Macherey-Nagel Bioanalysis™
740658.1 |
96 Preps |
239.00 €
96 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12765204
|
Macherey-Nagel Bioanalysis™
740658.2 |
192 Preps |
469.00 €
192 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12755214
|
Macherey-Nagel Bioanalysis™
740658.4 |
384 Preps |
887.00 €
384 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12745214
|
Macherey-Nagel Bioanalysis™
740658.24 |
2304 Preps |
3040.00 €
2304 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NucleoSpin™ 96 PCR Clean-up kit allows direct clean-up of PCR reaction mixtures. Within the procedure the addition of chaotropic salt (Buffer NT) allows a reversible adsorption of the PCR products to the silica membrane of the NucleoSpin™ 96 PCR Clean-up Binding Plates. High purity of the PCR-products is achieved by complete removal of primers, primer-dimers, salts, nucleotides, and proteins (e.g., polymerases, BSA) in subsequent washing steps using Buffer NT3. Highly pure PCR products are finally eluted with Elution Buffer NE (5 mM Tris/HCl, pH 8.5) or water (pH 8.5), and can be used directly for further applications.
- Silica membrane technology
- Processing: manual/automated, vacuum/centrifugation
- Sample material: < 10 0μL PCR reaction mixture
- Elution volume: 75 to 150 μL
- Binding capacity: 15 μg
- Preparation time: 45 min/plate
- Fragment size: 65 bp to 10 kb
Specifications
18°C to 25°C | |
96-well Plate | |
PCR clean up | |
< 100 μL |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.