missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Blood Column

Purify genomic DNA quickly and easily. NucleoSpin™ Blood Columns enable purification of genomic DNA from blood and biological fluids

Brand:  Macherey-Nagel™ 740951.50


 View more versions of this product

Product Code. 11942402

  • 188.00 € / 50 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Features
  • For rapid purification of high-quality DNA from blood
  • Ready-to-use DNA in less than 30 min
  • Complete removal of PCR inhibitors
  • Suitable for whole blood, treated with citrate, EDTA, heparin, CPDA
  • Consistently high DNA yields
  • Suitable for isolation of viral DNA
Applications
  • Genomic, bacterial, viral DNA isolation
  • DNA from whole blood (human or animal blood, fresh or frozen)
  • DNA from whole blood treated with citrate, EDTA, heparin, CPDA
  • DNA from serum, plasma, buffy coat, platelets, body fluids (e.g. amniotic fluid)
  • DNA from cultured cells
  • DNA suitable for (PCR, Southern blotting, enzymatic reactions, etc.)
Specifications

Specifications

Silica-membrane Technology
Silica Membrane Technology
For Genomic, Bacterial, Viral DNA Isolation
50
1 Each
4 to 6μg
Mini Spin Column
PCR, Southern Blotting, Enzymatic Reactions
2mL Collection Tubes, Buffers, Proteinase K
NucleoSpin™ Blood Column
30min.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash with water thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride / 2-propanol

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers