missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Blood L
Complete kit with silica membrane midi spin columns for rapid medium-scale purification of genomic DNA from blood and biological samples.
Brand: Macherey-Nagel™ 740954.24
This item is not returnable.
View return policy
Description
NucleoSpin™ Blood L
- Ready-to-use DNA in less than 60 min
- Complete removal of PCR inhibitors
- Consistently high DNA yields
- Suitable for whole blood (human or animal blood, fresh or frozen)
- DNA from whole blood treated with citrate, EDTA, heparin, CPDA
- DNA from serum, plasma, buffy coat, platelets, body fluids (e.g., amniotic fluid)
- DNA from cultured cells
- Typical downstream applications: PCR, Southern blotting, enzymatic reactions
Specifications
White band | |
Columns: 4 °C; Other components: Room temperature (18-25 °C) | |
Silica Membrane Technology | |
Binding capacity: 250 μg | |
Genomic DNA | |
Medium scale purification of DNA from blood | |
24 Preps | |
0.2 - 2 mL blood, 2 x 107 cells | |
40 - 60 μg (2 mL whole blood) |
200 to 500 ng/μL | |
Midi Spin Column | |
Cell Cultures, Whole Blood | |
Elution volume: 120 - 200 μL | |
Centrifuge | |
NucleoSpin Blood L Columns with Collection Tubes, Collection Tubes (15 mL), buffers, Proteinase K | |
A260/A280: 1.6 - 1.9 | |
60 min/prep |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Proteinase k, propan-2-ol, guanidine hydrochloride, ethanol