Learn More
Macherey-Nagel™ NucleoSpin Dx RNA Blood, IVD Kit
NucleoSpin Dx RNA Blood (IVD) for RNA isolation S-Monovette RNA Exact Isolation of RNA from the SARSTEDT's S-Monovette RNA Exact.
Brand:  Macherey-Nagel™ 740201.50
Product Code. 18031091
  • 559.00 € / Each
Explore more special offers
Description
Description
  • The NucleoSpin Dx RNA Blood kit is a IVD mini spin kit for the isolation of human RNA from whole blood collected in SARSTEDT's S-Monovette™ RNA Exact for the subsequent in-vitro diagnostic analysis (compliant with IVDR 2017/746).
  • The product provides purified human RNA to be used for subsequent down-stream analysis such as RT-PCR, qRT PCR, or RNA-Seq to obtain information about the RNA expression level in the sample.
  • The product is used by professional users in diagnostic laboratories.
Specifications
Specifications
50 Preps, 15 to 25 °C / 59 to 77 °F
Silica Membrane Technology
Whole Blood (Specific)
Isolation of RNA
NucleoSpin
1.6 to 2.2 (typically 1.9 to 2.1)
1.2 mL solution of stabilized blood from S-Monovette™ RNA Exact blood collection tubes (SARSTEDT REF 01.2048.001)
55 min./6 Preps
Mini prep
60 μL or 40 μL
Binding capacity: 200 μg
50 Preps
IVD Kit
50 Preps
RNA
>1 μg (0.7 to 4.2 μg) per preparation from blood from healthy subjects