missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Food Column

Isolate genomic DNA from food and feed. NucleoSpin™ Food Columns are perfect for small-scale DNA isolation for a variety of downstream applications.

52.50 € - 1060.00 €

Specifications

Detection Method Silica-membrane Technology
Format Mini Spin Column
For Use With (Equipment) PCR, Southern Blotting, Enzymatic Reactions
Includes 2mL Collection Tubes, Buffers, Proteinase K
Product Type DNA Isolation Columns
View More Specs

Products 3
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
11952512
View accessory products View Documents
Macherey-Nagel™
740945.10
10 Preps
52.50 €
10 reactions
Please to purchase this item. Need a web account? Register with us today!
11922272
View accessory products View Documents
Macherey-Nagel™
740945.250
250 Preps
1060.00 €
250 reactions
Please to purchase this item. Need a web account? Register with us today!
11972402
View accessory products View Documents
Macherey-Nagel™
740945.50
50 Preps
241.00 €
50 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Features
  • For rapid small-scale isolation of genomic DNA from food and feed
  • Complete removal of PCR inhibitors – get high quality DNA
  • Even low amounts of partially degraded DNA can be purified from complex matrices
  • Fast and easy procedure DNA from various sample materials – highest flexibility
Applications
  • DNA from complex matrices, e.g., processed food, soy (milk and flour), chocolate, cereals, meat, animal feed
  • Typical downstream applications: real-time PCR, Southern blotting, enzymatic reactions
Specifications

Specifications

Silica-membrane Technology
PCR, Southern Blotting, Enzymatic Reactions
DNA Isolation Columns
0.1 to 10μg
Mini Spin Column
2mL Collection Tubes, Buffers, Proteinase K
1 Each
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride/dodecyl sulfate,sodium salt/ethanol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.