missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ gDNA Clean-up XS, Micro kit for DNA clean up and concentration
Click to view available options
No. of Reactions:
10
50
Unit Size:
10 reactions
Each
Specifications
Specifications
| Detection Method | Silica-membrane Technology |
| Format | Mini Spin Column - XS Design |
| For Use With (Application) | Small DNA Quantities Extracted |
| Includes | 2mL Collection Tubes, Buffers |
| No. of Reactions | 50 |
| Product Type | NucleoSpin™ gDNA Clean-up XS Column |
| Quantity | 1 Each |
| Sample Volume | <400 μL solution containing <2 μg DNA |
| Test Time | 20min. |
| Yield | 60 to 70% |
Safety and Handling
Product Identifier
- NucleoSpin gDNA Clean-up XS (50)
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- MIXTURE LIST-Contains : guanidium thiocyanate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction