missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up XS
Concentrated clean up with small elution volumes from PCR reaction mixtures and agarose gels
39.60 € - 555.00 €
Specifications
Capacity | 5 μg |
---|---|
Column Type | XS Spin Column |
Content And Storage | Non hazardous, 18°C to 25°C |
Detection Method | Silica Membrane Technology |
Isolation Technology | Silica Membrane Technology |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
17192796
|
Macherey-Nagel™
740611.10 |
10 |
39.60 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17112806
|
Macherey-Nagel™
740611.50 |
50 |
137.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17102806
|
Macherey-Nagel™
740611.250 |
250 |
555.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Rapid concentration and clean up of PCR products
- 2 in 1 kit for reaction mixtures and gel extraction
- XS column design enables elution of highly concentrated DNA
Specifications
5 μg | |
Non hazardous, 18°C to 25°C | |
Silica Membrane Technology | |
6 to 12 μL | |
Agarose gel | |
No | |
pH-Indicator included | |
DNA | |
A260/A280: 1.8-1.9 | |
Room Temperature | |
DNA Fragments | |
Recovery 70-95% |
XS Spin Column | |
Silica Membrane Technology | |
Agarose Gel, PCR Reaction Mixture | |
Centrifugation, Vacuum, Vortex Mixer | |
PCR Clean-up and Gel Extraction, Sample Concentration | |
NucleoSpin Gel and PCR Clean-up XS Columns, Collection Tubes, buffers | |
Gel extraction and PCR clean-up | |
Nucleic Acid Purification Kit | |
<200 μL PCR reaction mixture, < 200 mg agarose gel | |
50 bp-20 kbp | |
15 min / 6 preps |
Safety and Handling
missing translation for 'shelfLife' : 24 months
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Supplemental information
- NucleoSpin Gel and PCR Clean-up XS (10)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Supplemental information
- MIXTURE LIST-Contains : guanidium thiocyanate
For research use only