missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Plant II
Isolate and purify genomic DNA from plant samples. NucleoSpin™ Plant II kits enable enhanced yield and quality DNA.
51.70 € - 1010.00 €
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11992492
|
Macherey-Nagel™
740770.10 |
10 Preps |
51.70 €
10 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11912262
|
Macherey-Nagel™
740770.250 |
250 Preps |
1010.00 €
250 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11982382
|
Macherey-Nagel™
740770.50 |
50 Preps |
228.00 €
50 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Features
- The second generation for rapid isolation of genomic DNA from plant samples
- with higher yield and quality
- Two alternative lysis buffers included for optimal processing of various samples and highest flexibility:
- Lysis Buffer PL1, based on CTAB lysis method
- Lysis Buffer PL2, based on SDS lysis method
- NucleoSpin™ Filters included for clarification of lysate
- Improved buffer compositions – higher yield and purity
- Optimized silica membrane – improved DNA binding
- RNase A included
Applications
- DNA from plant cells and tissue
- DNA from fungi
- Typical downstream applications: PCR, Southern blotting, enzymatic reactions
Specifications
Plant II |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : guanidine hydrochloride, ethanol/dodecylsulfate, sodium salt/guanidine hydrochloride, 2-propanol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title