missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Columns, Mini columns for RNA purification

Product Code. 15870968
Click to view available options
Quantity:
250 Columns
50 Columns
Unit Size:
Pack of 250
Pack of 50
This item is not returnable. View return policy

Product Code. 15870968

Brand: Macherey-Nagel™ 740955.250S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Additional silica-membrane mini spin columns for use in isolating RNA of the highest integrity from cells and tissue using the NucleoSpin™ RNA kit.

NucleoSpin™ RNA

  • rDNase included for on-column DNA digestion– easy and efficient removal of contaminating DNA
  • NucleoSpin Filters (shredders) – efficient sample homogenization and reduction of viscosity
  • Up to 70 μg ready to use RNA
  • Parallel purification of genomic DNA by using the NucleoSpin™ RNA/DNA Buffer Set
  • RNA isolation from cultured cells, tissue (standard protocol):
    • RNA from < 109 bacterial cells (Gram-negative, Gram-positive) or < 108 yeast cells
    • RNA from ≤ 100 μL biological fluids
    • RNA clean-up from reaction mixtures
    • RNA from samples stored in RNAlater
    • RNA from saliva samples collected with Oragene™ RNA (Genotek)
  • Typical downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays

Specifications

Color Blue band
Content And Storage rDNase at 4 °C; Other components at room temperature (18°C to 25 °C)
Format Mini Spin Column
Isolation Technology Silica Membrane Technology
Elution Volume 40 to 120 μL
Sample Type Cultured Bacterial or Yeast Cells, Human or Animal Tissue
Efficiency Binding capacity: 200 μg
For Use With (Equipment) NucleoSpin RNA Kits
For Use With (Application) Isolation of RNA from cells and tissue
Includes Columns only
Quantity 250 Columns
Sample Size < 30 mg tissue, 5 x 106 cultured cells
Test Time 35 min/6 preps
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin RNA
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate, ethanol (diluted < 20 %) (denaturedwith 1%IPA/1%MEK, acc.2016/1867/EU); gu
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.