missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ RNA, Mini kit for RNA purification
Click to view available options
No. of Reactions:
10
12
50
240
250
Format:
Mini Spin Column
QIAcube
Unit Size:
10 reactions
250 reactions
300mL
50 reactions
Each
Descripción
Features
- DNase included for on-column DNA digestion– easy and efficient removal of contaminating DNA
- NucleoSpin™ Filters (shredders) – efficient sample homogenization and reduction of viscosity
- Up to 70 μg ready-to-use RNA
- Parallel purification of genomic DNA by using the NucleoSpin™ RNA/DNA Buffer Set
- RNA isolation from cultured cells, tissue (standard protocol)
- RNA from < 109 bacterial cells (Gram-negative, Gram-positive) or < 108 yeast cells
- RNA from ≤ 100 μL biological fluids
- RNA clean-up from reaction mixtures
- RNA from samples stored in RNAlater™
- RNA from saliva samples collected with Oragene™ RNA (Genotek)
- Typical downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays
Especificaciones
Especificaciones
| Bacterial Content | 70 μg |
| Detection Method | Silica-membrane |
| Format | Mini Spin Column |
| Isolation Technology | Silica Membrane |
| DNase | Yes |
| Final Product Type | NucleoSpin™ RNA Kit |
| For Use With (Application) | RNA isolation from cultured cells, tissue |
| No. of Reactions | 50 |
| No. of Samples | 4 |
| Product Type | NucleoSpin™ RNA Kit |
| Mostrar más |
Seguridad y manipulación
Palabra de advertencia
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : guanidinium thiocyanate, ethanol (diluted < 20 %) / guanidine hydrochloride, ethanol
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido