missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA, Mini Kit for RNA purification

Mini spin kit for isolation of RNA of highest integrity.

Brand:  Macherey-Nagel™ 740955.12C

70.92 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.



 View more versions of this product

Product Code. 15373604

  • 76.55 € / 300mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Features
  • DNase included for on-column DNA digestion– easy and efficient removal of contaminating DNA
  • NucleoSpin™ Filters (shredders) – efficient sample homogenization and reduction of viscosity
  • Up to 70 μg ready-to-use RNA
  • Parallel purification of genomic DNA by using the NucleoSpin™ RNA/DNA Buffer Set
Applications
  • RNA isolation from cultured cells, tissue (standard protocol)
  • RNA from < 109 bacterial cells (Gram-negative, Gram-positive) or < 108 yeast cells
  • RNA from ≤ 100 μL biological fluids
  • RNA clean-up from reaction mixtures
  • RNA from samples stored in RNAlater™
  • RNA from saliva samples collected with Oragene™ RNA (Genotek)
  • Typical downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays
Specifications

Specifications

70 μg
QIAcube
Yes
RNA isolation from cultured cells, tissue
NucleoSpin™ RNA Kit
Silica-membrane
Silica Membrane
NucleoSpin™ RNA Kit
12
1 Each
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • NucleoSpin RNA

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate, ethanol (diluted < 20 %) (denaturedwith 1%IPA/1%MEK, acc.2016/1867/EU); gu

SDS
Documents

Documents

Certificates
Special Offers

Special Offers