missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Plant Kit

Isolates total RNA from all kinds of plant material and filamentous fungi without degradation of the RNA. Macherey-Nagel™ NucleoSpin™ RNA Plant Kit is a complete mini spin kit that includes RNA plant columns with collection tubes, filters, all needed buffers and RNase-free rDNase.

76.90 € - 1655.00 €

Specifications

Format Mini Spin Column
For Use With (Equipment) Real-time RT-PCR
For Use With (Application) RNA purification from plant cells and tissue
Product Type NucleoSpin™ RNA Plant Column
Quantity 1 Each
View More Specs

Products 3
Product Code Brand No. of Reactions Includes Price Quantity & Availability  
Product Code Brand No. of Reactions Includes Price Quantity & Availability  
12979894
View Documents
Macherey-Nagel™
740949.10
10 Preps Lysis buffer RA1 (10mL), Lysis buffer RAP (10mL), Wash buffer RAW2 (13mL), Wash Buffer RA3 concentrate (6mL), Membrane desalting buffer (10mL), Reaction buffer for rDNase (7mL), RNase-free rDNase (1 size D vial), RNase-free water (13mL), NucleoSpin filters (10), NucleoSpin RNA Plan Columns with collection tubes (10), 2mL Collection tubes (30), 1.5mL Collection tubes (10)
76.90 €
10 reactions
Please to purchase this item. Need a web account? Register with us today!
12721021
View accessory products View Documents
Macherey-Nagel™
740949.50
50 Preps Lysis buffer RA1 (25mL), Lysis buffer RAP (25mL), Wash buffer RAW2 (13mL), Wash Buffer RA3 concentrate (12mL), Membrane desalting buffer (25mL), Reaction buffer for rDNase (7mL), RNase-free rDNase (1 size F vial), RNase-free water (13mL), NucleoSpin filters (50), NucleoSpin RNA Plan Columns with collection tubes (50), 2mL Collection tubes (150), 1.5mL Collection tubes (50)
380.00 €
50 reactions
Please to purchase this item. Need a web account? Register with us today!
12711021
View accessory products View Documents
Macherey-Nagel™
740949.250
250 Preps Lysis buffer RA1 (125mL), Lysis buffer RAP (125mL), Wash buffer RAW2 (80mL), Wash Buffer RA3 concentrate (3 x 25mL), Membrane desalting buffer (125mL), Reaction buffer for rDNase (30mL), RNase-free rDNase (5 size F vials), RNase-free water (60mL), NucleoSpin filters (250), NucleoSpin RNA Plan Columns with collection tubes (250), 2mL Collection tubes (750), 1.5mL Collection tubes (250)
1655.00 €
250 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NucleoSpin™ RNA Plant Kit is designed for isolation of DNA-free, high-quality RNA from a wide variety of plant and fungal samples. Typical downstream applications include real-time RT-PCR, gene expression profiling, Northern blotting, primer extension, array technology, and RNase protection assays.

  • Two different lysis buffers included—Optimized lysis procedure for typical conditions and for peculiar metabolites
  • rDNase included for on-column digestion—Efficient removal of contaminating DNA
  • NucleoSpin™ Filters (shredders) included—Efficient sample homogenization and reduction of viscosity
  • Parallel purification of DNA possible—By using the NucleoSpin™ RNA/DNA buffer set

Kit Components:
NucleoSpin™ RNA columns with 2mL collection tubes, 2mL and 1.5mL collecting tubes, NucleoSpin™ filters, buffers (lysis, wash, desalting), RNase-free DNase I, reaction buffer for DNase, RNase-free water

  • Mini spin column with silica membrane technology
  • Yields 3–70μg RNA from 100μg plant material
  • Elution volume: 40–60μL
  • Fragment size: 200b–20kb
  • Binding capacity: 200μg
  • Preparation time: 30 min/6 preps
  • A260/A280: 1.9–2.1
Specifications

Specifications

Mini Spin Column
RNA purification from plant cells and tissue
1 Each
3–70μg
Real-time RT-PCR
NucleoSpin™ RNA Plant Column
<100mg tissue
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : ethanol (diluted < 20 %)(denatured with1%IPA/1%MEK, acc.2016/1867/EU); guanidinium thiocyanate; gua

SDS
Documents

Documents

Product Certifications