missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Plus Kit

Efficient and quick RNA isolation from cultured cells, animal tissue, plant tissue, microorganisms. Macherey-Nagel™ NucleoSpin™ RNA Plus Kit includes the NucleoSpin™ gDNA Removal Column, a spin column, which quickly and effectively removes genomic DNA contamination without DNase digestion.

Brand:  Macherey-Nagel™ 740984.10

69.80 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.



Product Code. 15370195

  • 82.55 € / 10 preps

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

During the isolation of RNA, it is imperative to prevent the degradation of the RNA. In the NucleoSpin™ RNA Plus method, cells and tissues are first lysed by incubation in a chaotropic ion lysis buffer solution, which immediately inactivates RNases. The lysate is added to the gDNA Removal Column to clarify the lysate and to remove contaminating gDNA. After the addition of the Binding Solution to the flow-through, the RNA is bound to the RNA Plus Column. Two subsequent wash steps remove salts, metabolites, and macromolecular cellular components. High quality RNA is eluted with RNase-free H2O.

  • Includes NucleoSpin™ gDNA Removal Column: no time consuming rDNase digestion
  • Convenient handling: lysate clearing and gDNA removal with one column in one step
  • New efficient lysis buffer: no β-mercaptoethanol or TCEP necessary
  • Downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays
  • Kit contents: NucleoSpin RNA Plus Columns, NucleoSpin gDNA Removal Columns, Collection Tubes, buffers
Specifications

Specifications

<109 Cells
Lysis Buffer
Silica Membrane System (Two Columns)
<1 x 107 Cultured Cells, <1 x 109 Bacterial Cells, <1 x 108 Yeast Cells, <30 mg Human/Animal Tissue
NucleoSpin™ RNA Plus Columns with Collection Tubes, NucleoSpin™ gDNA Removal Columns with Collection Tubes, Collection Tubes (2 mL), Collection Tubes (1.5 mL), buffers
NucleoSpin RNA Plus kit
Yes
20 min./6 Reaction
Room temperature (18-25°C)
Mini Spin Column
Elution volume: 30–120μL
RNA isolation from cultured cells, animal tissue, plant tissue, microorganisms
10
1 Kit
Fragment size: >200 nt
HeLa cells 5 x 106: 40-60μg
Mouse liver 20mg: 80-100μg
Mouse kidney 20mg: 40-70μg
Mouse spleen 5mg: 30-60μg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

Supplemental information
  • MIXTURE LIST-Contains : ethyleneglycol diacetate / guanidinium thiocyanate /guanidine hydrochloride, ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers