missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ RNA Plus XS
Designed for efficient and quick RNA isolation from the smallest samples of cells and tissues. State-of-the-art, dual-column kit includes gDNA removal columns and RNA binding columns.
Brand: Macherey-Nagel™ 740990.250
This item is not returnable.
View return policy
Description
NucleoSpin™ RNA Plus XS
- Developed for extra small samples down to single cell analysis
- Efficient and quick RNA isolation from cells and tissues
- Two-column system includes gDNA removal columns and RNA binding columns
- NucleoSpin™ gDNA Removal Columns – no time consuming rDNase digestion
- Convenient handling – lysate clearing and gDNA removal on one column in one step
- New efficient lysis buffer – no b-mercaptoethanol or TCEP necessary
- Variety of sample materials – cultured cells, animal tissue, plant tissue, microorganisms
- Downstream applications include real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays
Specifications
Yellow/Blue ring | |
Mini Spin Column | |
5 - 30 μL | |
Binding capacity: 110 μg | |
Centrifuge | |
NucleoSpin RNA Plus XS Columns, NucleoSpin gDNA Removal Columns XS, Collection Tubes (2 mL), Collection Tubes (1.5 mL), Buffers | |
RNA Isolation Kit | |
1 - 105 cultured cells; < 5 mg human / animal tissue | |
HeLa cells (105): 500 - 2000 ng Mouse liver (0.5 μg): 2.5 - 8 ng Mouse brain (0.5 μg): 0.1 - 0.5 ng |
Room temperature (18-25 °C) | |
Silica Membrane | |
Cultured Cells, Human or Animal Tissue | |
RNA (with typical RIN > 8) | |
Isolation of RNA | |
250 Preps | |
A260/A280: 1.9 - 2.2 | |
18 min/6 preps |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Supplemental information
- NucleoSpin RNA Plus XS
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanate/guanidinium thiocyanate,ethanol (diluted < 20 %)