missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA/Protein, Mini kit for RNA and Protein Purification

Product Code. 11912382
Click to view available options
Sufficient For:
10rxns
250rxns
50rxns
Unit Size:
10 reactions
250 reactions
50 reactions
This item is not returnable. View return policy

Product Code. 11912382

Brand: Macherey-Nagel™ 740933.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate RNA and protein from undivided samples. Macherey Nagel™ NucleoSpin™ RNA/Protein Column Set solves the problem of extracting both protein and RNA from the same sample in parallel.

Features
  • Parallel isolation of RNA and protein from undivided samples
  • No splitting of samples required – reliable interpretation of RNA and protein extracted from the same sample (direct correlation)
  • High RNA yield and integrity
  • High protein yield independent of protein size, localization, modification, etc.
  • Easy protein quantification using the MACHEREY-NAGEL Protein Quantification Assay
  • Complete mini kit with NucleoSpin™ Filters (shredders) and recombinant DNase for
  • DNA-free RNA of high quality
Applications
  • Rapid purification of total RNA and protein from cultured cells and tissue
  • Gene expression profiling, siRNA experiments, analysis of transgenic organisms, drug screening
  • Broad spectrum of starting material tested and proteins already detected

Specifications

Product Type NucleoSpin™ RNA/Protein Column Set
Quantity 1 Set
Includes 2mL Collection Tubes, 1.5mL Collection Tubes, NucleoSpin™ Filters, Buffers, RNase-free and rDNase
Sufficient For 50rxns
Column Type Spin Column
For Use With (Application) For Rapid Purification of Total RNA and Protein from Cultured Cells and Tissue
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate,ethanol(diluted< 20 %)/dodecyl sulfate, sodium salt/tris-(2-carboxyethyl)ph
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.