missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA XS Column

Purify high quality RNA quickly and easily. NucleoSpin™ RNA XS Columns enable purification of highly concentrated RNA from the smallest samples

Brand:  Macherey-Nagel™ 740902.250

Code : LE

Additional Details : Weight : 0.32000kg Transport : UN number : 1993 Chem class : 3 Pack group : III


 View more versions of this product

Product Code. 12713580

  • 1700.00 € / 250 reactions
Estimated Shipment: 30-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Features
  • Isolation of RNA from small sample quantities like biopsy material or single cells
  • Excellent RNA recovery and integrity
  • Concentrated RNA for sensitive downstream applications by elution in as little as 5 μL
  • rDNase included for on-column DNA removal
  • NucleoSpin™ Filters (shredders) – efficient homogenization and reduction of viscosity 
  • High quality RNA, ready to use for RT-PCR and other applications
  • Reducing agent TCEP – no ß-mercaptoethanol necessary
Applications
  • RNA isolation from cultured cells
  • RNA isolation from tissue
  • RNA isolation from cryosections
  • RNA isolation from laser captured cells
  • RNA isolation from small amounts of plant material
  • RNA isolation from samples stored in RNAlater™
  • Typical downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology,
  •  RNase protection assays
Specifications

Specifications

Mini Spin Column
For RNA isolation from cultured cells, tissue, cryosections and laser captured cells
250
1 Each
NucleoSpin™ RNA XS Column
Collection tubes, NucleoSpin™ filters, collection tubes (2mL), Collection tubes (1.5mL), buffers, RNase-free rDNase, carrier RNA, reducing agent TCEP
NucleoSpin™ RNA XS Column
0.1 to 1.5ng (102 HeLa cells), 10 to 15ng (103 HeLa cells), 100 to 150ng (104 HeLa cells), 1000 to 1500ng (105 HeLa cells)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate, ethanol (diluted < 20 %) (denaturedwith 1%IPA/1%MEK, acc.2016/1867/EU); tr

SDS
Documents

Documents

Certificates
Special Offers

Special Offers