missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Tissue
Mini spin kit for purification of DNA from a broad range of samples
46.50 € - 964.00 €
Specifications
Cell Type | Animal & Human tissue, cells |
---|---|
Capacity | 60 μg |
Column Type | Mini spin column |
Content And Storage | Harzadous, 18°C to 25°C |
Description | Kit for purification of DNA from a broad range of samples |
Alert:
UN 1993 Flammable liquid, n.o.s. (2-propanol mixture) Chem class:3 Pack Group III
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11902512
|
Macherey-Nagel™
740952.10 |
10 |
46.50 €
10 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10498961
|
Macherey-Nagel™
740952.50 |
50 |
212.00 €
50 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11982262
|
Macherey-Nagel™
740952.250 |
250 |
964.00 €
250 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Versatile kit for the isolation of genomic DNA from a wide range of different starting materials (cells, tissue, forensic & clinical samples)
- 16 support protocols available in the user manual for highest flexibility
- Highly sensitive membrane for reliable purification of even trace amounts of DNA
- Rapid and easy protocol (20 min/prep excl. lysis)
Specifications
Animal & Human tissue, cells | |
Mini spin column | |
Kit for purification of DNA from a broad range of samples | |
Animal and Human Tissue, Cells | |
DNA | |
Genotyping, Multiplex PCR, Next Generation Sequencing | |
DNA purification | |
1 Each | |
<25 mg human/animal tissue, 102-107 cells | |
200 bp to 50 kbp | |
DNA | |
20 min/prep (excl. lysis) |
60 μg | |
Harzadous, 18°C to 25°C | |
Silica Membrane Technology | |
60 to 100 μL | |
Centrifugation, Vacuum | |
NucleoSpin Tissue Columns, Collection Tubes, buffers, Proteinase K | |
DNA | |
A260/A280 1.7 - 1.9 | |
Room Temperature | |
Animal & Human tissue, cells | |
Silica-membrane technology |
Safety and Handling
missing translation for 'shelfLife' : 27 months after production
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : guanidine hydrochloride / 2-propanol
For research use only