missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Sample Processing & Storage Microplates
GREENER_CHOICE

Lower spending when storing compounds for small volume applications by using these 384-shallow well standard height polypropylene microplates.

Brand:  Thermo Scientific™ 267462

1071.00 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10742024

  • 1530.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Lower spending when storing compounds for small volume applications by using Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates. Polypropylene is ideal for storage plates because it has a lower binding capacity to prevent proteins or DNA binding, and allows for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors to allow for quick identification of storage plates.

Features:

  • Rounded wells are optimized for mixing and sample recovery
  • Square rounded wells minimize wicking (capillary action)
  • Natural plates for storage identification or storing colorimetric assays
  • White plates for luminescent assays
  • Black plates for fluorescent assays
  • Customized barcoding is available
TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Barcode
384-well
384-well Polypropylene Sample Processing & Storage Plates
Autoclavable
Low binding PP
White
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
55 μL
2 to 35 μL
Non-treated
Yes
100
For Research Use Only. Not for use in diagnostic procedures.
Certified Quality
Certified RNase and DNase Free. ANSI Compliant.
Without Lid
16 x 24
100 Cases
Nunc
384-well plate
Polypropylene
384
Non-sterile
Square, Round Bottom (U-well)
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
25
Yes
ANSI/ SLAS Standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers