missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Sample Processing & Storage Microplates
Lower spending when storing compounds for small volume applications by using these 384-shallow well standard height polypropylene microplates.
Brand: Thermo Scientific™ 267459
This item is not returnable.
View return policy
Description
Lower spending when storing compounds for small volume applications by using Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates. Polypropylene is ideal for storage plates because it has a lower binding capacity to prevent proteins or DNA binding, and allows for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors to allow for quick identification of storage plates.
Features:- Rounded wells are optimized for mixing and sample recovery
- Square rounded wells minimize wicking (capillary action)
- Natural plates for storage identification or storing colorimetric assays
- White plates for luminescent assays
- Black plates for fluorescent assays
- Customized bar coding is available
Specifications
No Barcode | |
384-well | |
384-well Polypropylene Sample Processing & Storage Plates | |
Autoclavable | |
Low binding PP | |
Natural | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
55 μL | |
2 to 35 μL | |
Non-treated | |
Yes | |
100 | |
100/Cs. | |
Yes | |
ANSI/ SLAS Standard |
Certified RNase and DNase Free. ANSI Compliant. | |
Without Lid | |
16 x 24 | |
25 pk., 100 cs. | |
Nunc | |
384-well plate | |
Polypropylene | |
384 | |
Non-sterile | |
Square, Round Bottom (U-well) | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
25 | |
For Research Use Only. Not for use in diagnostic procedures. | |
Certified Quality |