missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ IVF Multi Dish, 4-well

Perform applications which yield consistently reliable, high-quality results during vitrification.

Brand:  Thermo Scientific™ 179830

 View more versions of this product

Product Code. 10756864

  • 441.00 € / Pack of 120

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Generate consistently reliable results during IVF and embryo vitrification with Thermo Scientific™ Nunc™ 4-Well IVF Dishes, which meet the rigorous requirements of IVF testing, certification and validation.

Innovative features for high quality and ease-of-use:

  • Ensures proper well orientation and prevents cross contamination with notched lid and dish
  • Prevents scratches on well bottoms and promotes fast temperature equilibrium with 0.8mm raised edge
  • The Nunclon Delta surface treatment ensures consistency of a more hydrophilic surface
  • The non-treated hydrophobic surface facilitates microdrop embryo culture
  • Certificates of conformity available for every lot

Rigorously tested, certified and validated for IVF:

  • CE-marked (Notified Body 2460), FDA-listed
  • Produced under ISO 13485 with full lot traceability
  • Sterile (SAL 10-6)
  • Non-pyrogenic (LAL tested)
  • USP Class VI polystyrene
  • Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)

Warranty: 90 days

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested
1.9 cm2/well
Sterile
4
4
66 mm
Non-treated IVF Dish, 4-well
Polystyrene (USP Class VI)
Non-treated
120
66 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.