missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ 50 mL Bioreactor Tube

Thermo Scientific™ Nunc™ Bioreactor Centrifuge Tubes are designed for high throughput protein expression screening and cell line development and optimization under suspension condition.

Brand:  Thermo Scientific™ 332260

66.00 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 30054604

  • 110.00 € / Pack of 90

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Nunc™ Bioreactor Centrifuge Tubes are designed for high throughput protein expression screening and cell line development and optimization under suspension condition. Adjustable 4-level vented cap with a 0.2 μm filter membrane allows for controlled gas exchange and air circulation without contamination. Ergonomic grooved cap for effortless handling and a large writing area for ease of identification.

  • Ideal for screening, small volume high throughput analysis
  • Protein expression suspension culture volumes ranging from 10 mL to 25 mL
  • Convenient pack sizes
  • Four different sizes of ventilation (dial type) on the cap.
  • Tube: Polypropylene
  • Cap: HDPE
  • Filter: PTFE
  • Dial: Polystyrene
  • 2 μm PTFE filter membrane
  • Suitable for suspension culture
  • Sterile - SAL 10-6
  • Non-pyrogenic
  • Non-cytotoxic
  • Shelf life: 2 years
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HDPE
Single-use
Ideal for screening, small volume high throughput analysis
90
Sterile
Yes
Conical
Yes
Protein expression suspension culture volumes ranging from 10 mL to 25 mL
Polypropylene
  • Cap: HDPE
  • Filter: PTFE
  • Dial: Polystyrene 
  • 0.2 μm PTFE filter membrane
3
Yes
Autoclavable
Quadruple-position Vent/Close Cap
Conical Centrifuge Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers