missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ 96 DeepWell™ Polystyrene Plates
Clone and replicate assays with these untreated polystyrene plates, suited for agglutination assays and sample collection.
237.00 € - 289.00 €
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
Packaging | 32 Cases, 4 Pk. |
Product Line | Nunc |
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
10577131
|
Thermo Scientific™
278605 |
Non-sterile |
237.00 €
Pack of 32 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
10070681
|
Thermo Scientific™
278606 |
Sterile |
289.00 €
Pack of 32 |
In Stock
Log in to see stock. |
|||||
Description
Store samples and perform agglutination assays with Thermo Scientific™ Nunc™ 96 DeepWell™ Polystyrene Plates, featuring transparent round bottom deep wells that hold up to 900μL per well.
- Alpha-numeric grid provides quick and easy sample identification
- Optimized for robotics and liquid handling systems
- Round-bottom wells for optimal sample recovery
- Non-treated hydrophobic surface
- Sealable with well caps (Part No. 278616) or sealing tape
- Customized bar coding is available
Specifications
Standard | |
8 x 12 | |
Nunc | |
Microplate | |
96 | |
Non-treated | |
32 | |
128 x 86 x 41 mm |
Without Lid | |
32 Cases, 4 Pk. | |
Clear | |
Polystyrene | |
900 μL | |
Round Bottom (U-well) | |
4 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.