missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
GREENER_CHOICE

Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.

Brand:  Thermo Scientific™ 249944

216.52 EUR valid until 2025-05-31
Use promo code "25075" to get your promotional price.


 View more versions of this product

Product Code. 10525511

  • 299.00 € / Pack of 120
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.

Features:

  • Shared-wall design increases the well volume
  • Wider wells and rounded bottom improve mixing
  • Conical bottom well for optimal sample recovery
  • Black and white plates available for fluorescent- and luminescent-based applications
  • Low binding polypropylene for homogeneous assays and storage
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1425596 Certificate TypeCertificate of Compliance Date29/03/2025 Catalog Number249944
Lot Number1425112 Certificate TypeCertificate of Compliance Date27/03/2025 Catalog Number249944
Lot Number1423761 Certificate TypeCertificate of Compliance Date25/03/2025 Catalog Number249944
Lot Number1422643 Certificate TypeCertificate of Compliance Date12/02/2025 Catalog Number249944
Lot Number1422541 Certificate TypeCertificate of Compliance Date31/01/2025 Catalog Number249944
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates > 96 Conical Well, Natural Polypropylene, Non-Treated, without lid, Nonsterile

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.