Learn More
Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.
Brand: Thermo Scientific™ 95040410
Description
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
- Shared wall design increase the well volume
- Wider wells and rounded bottom improve mixing
- Conical bottom well are for optimal sample recovery
- Excellent chemical resistance
- Withstand temperatures from -80° to +121°C (-112° to +250°F)
- Black and white plates are available for fluorescent- and luminescent-based applications
- Low binding polypropylene for homogeneous assays and storage

Specifications
No Barcode | |
Standard | |
96-well Polypropylene Sample Processing & Storage Plates | |
Not Autoclavable | |
Low binding PP | |
Natural | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
500 μL | |
Non-sterile | |
Square, Conical Bottom (V-well) | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
25 | |
Yes | |
ANSI/ SLAS Standard |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
Without Lid | |
8 x 12 | |
50 Cases | |
Nunc | |
96-well plate | |
Polypropylene | |
96 | |
Non-treated | |
Yes | |
50 | |
For Research Use Only. Not for use in diagnostic procedures. | |
Certified Quality |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.