Learn More
Thermo Scientific™ Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
BIOPHARMA
Recover samples completely with these conical well bottom plates. The bottom shape allows for easy pipette access.
Brand:  Thermo Scientific™ 249570
240.77 EUR valid until 2024-12-31
Use promo code "21894" to get your promotional price.
 View more versions of this product
Product Code. 10580382
  • 321.00 € / Pack of 180
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Recover samples completely with Thermo Scientific™ Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates. The bottom shape allows for easy pipette access. The plates are optimized for robotics, automated readers and liquid handling systems.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
TRUSTED_SUSTAINABILITY
Specifications
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Without Lid
180 Cases
Microplate
96
Non-treated
180
180/Cs.
Standard
8 x 12
Clear
Polystyrene
Non-sterile
Conical Bottom (V-well)
10
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.