Learn More
Thermo Scientific™ Nunc™ Cell Factory™ Systems
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems.
233.28 € - 1660.00 €
Product Code | Brand | Culture Area | Tray Layers | No. per Case | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Culture Area | Tray Layers | No. per Case | Price | Quantity & Availability | |||
10344321
|
Thermo Scientific™
139446 |
25,280 cm2 | 40 | 2 |
1660.00 €
Pack of 2 |
Estimated Shipment: 24-04-2025
Log in to see stock. |
|||
10113922
|
Thermo Scientific™
140004 |
2528 cm² | 4 | 10 |
1245.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||
12677093
|
Thermo Scientific™
140004 |
2528 cm² | 4 | 10 | N/A | ||||
10049040
|
Thermo Scientific™
164327 |
6320 cm² | 10 | 2 |
442.00 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
10634551
|
Thermo Scientific™
165250 |
632 cm² | 1 | 8 |
476.00 €
Pack of 8 |
In Stock
Log in to see stock. |
|||
10394221
|
Thermo Scientific™
167695 |
1264 cm² | 2 | 6 |
381.00 €
Pack of 6 |
In Stock
Log in to see stock. |
|||
13101594
|
Thermo Scientific™
167695 |
1264 cm² | 2 | 6 | N/A | ||||
10604901
|
Thermo Scientific™
170009 |
6320 cm² | 10 | 6 |
1225.00 €
Pack of 6 |
In Stock
Log in to see stock. |
|||
Description
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems. A proven solution for large-scale production of cells, vaccines and therapeutic proteins, these systems have the same growth kinetics as laboratory-scale cell culture product. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling and harvesting. Available in 1-, 2-, 4-, 10- and 40-tray versions.
- Ideal for adherent cells
- Boost production efficiency with more surface area in a small footprint
- Reduce contamination risks with a closed system
Specifications
Original Cell Factory | |
Cell Factory System |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.