missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ Containers
Store samples safely using Thermo Scientific™ Nunc™ Containers.
Brand: Thermo Scientific™ 536080
213.56 EUR valid until 2025-12-31
Use promo code "24277" to get your promotional price.
This item is not returnable.
View return policy
Description
Store samples safely using Thermo Scientific™ Nunc™ Containers. Nunc Labware Products are made from high purity resins and molded using our state-of-the-art processes. Plastic labware is a safer alternative to glass without sacrificing accuracy.
Features:- Transparent polystyrene construction
- Versatile containers made of high-quality resins
- Snap caps available for all containers (not included)
- Nonsterile (except for 561301)

Specifications
Container | |
Polystyrene | |
Non-sterile | |
Snap Cap |
No | |
Cylindrical | |
12.5 mL | |
Clear |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Nunc™ Containers >
Spot an opportunity for improvement?Share a Content Correction