Learn More
Thermo Scientific™ Nunc™ Disposable Loops and Needles, needle
Product Code. 10508731
Click to view available options
:
Needle; Carton of 1000
Product Code. 10508731
Brand: Thermo Scientific™ 253988
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
  • Loops available in 10 and 1μL sizes
  • Meet semi-quantitative standard for handling samples
  • Calibration documents available for any batch upon request
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Material Polystyrene
Sterility Sterile
Color Yellow
Product Type Inoculating Needle
Quantity Carton of 1000