missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate Product Code.: 15543115
Add to basket

Thermo Scientific™ Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Product Code. 15543115
160 Microplates
Click to view available options
Quantity:
160 Microplates
50 Microplates
This item is not returnable. View return policy

Product Code. 15543115

Brand: Thermo Scientific™ 167542

Add to basket
Add to basket
This item is not returnable. View return policy

The Thermo Scientific™ Nunc™ Edge 2.0 Plate provides an efficient solution for preventing evaporation and in turn the risks associated with edge effect.

Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.

Features
  • The Edge 2.0 Plate provides an efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect.
  • The surrounding built-in moat is divided into 4 sectional reservoirs, each can be filled with 1.7 to 2.0mL sterile water, buffer or media.
  • Replenish fluid in the reservoirs every 3 days in the humidified incubator to ensure the effectiveness of the evaporation barrier.
  • Nunclon™ Delta cell culture surface treatment of the wells promotes cell attachment and growth.
  • The non-treated surface is available for suspension cell culture or general solution-based assays
  • Use the Edge 2.0 Plate in conjunction with the Thermo Scientific™ Varioskan™ LUX multimode microplate reader. The filled moat of the plate functions as passive humidity control, allowing for simultaneous incubation and signal detection during long-term applications with living cells inside the plate reader.
Warranty: 90 days
Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
    • Renewable Materials
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Certified Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
Lid With Lid
Culture Area 0.35 cm2/well
Array 8 x 12
Packaging 10 Pk., 160 Cases
Product Line Nunc
Color Clear
Product Type Microplate
Skirt Style Low Flange
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Nunclon Delta
Well Shape Flat Bottom (F-well)
Green Features Less waste, fewer resources used
No. per Case 160
No. per Pack 10
Quantity 160 Microplates
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.