Learn More
Thermo Scientific™ Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate

Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate.
Brand: Thermo Scientific™ 167574
124.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.

Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
Without Lid | |
8 x 12 | |
Nunc | |
Microplate | |
Polystyrene | |
96 | |
Nunclon Delta | |
Less waste, fewer resources used | |
1 |
Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility | |
0.35 cm2/well | |
1/Pk., 50/Cs. | |
Clear | |
Low Flange | |
400 μL | |
Sterile | |
Flat Bottom (F-well) | |
50 | |
Case of 50 |
Videos
-
Thermo Scientific Nunc Edge 2.0 plate
-
Thermo Scientific Nunc Edge 2.0 plate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
185261 | Certificate of Irradiation | 15/04/2025 | 167574 |
185261 | Certificate of Compliance | 06/03/2025 | 167574 |
182422 | Certificate of Compliance | 03/12/2024 | 167574 |
175389 | Certificate of Compliance | 03/12/2024 | 167574 |
180217 | Certificate of Compliance | 03/12/2024 | 167574 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.