Learn More
Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes
Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes can be opened and closed with one hand due to a proprietary hinged-cap system. The tubes feature an ergonomic design, and are compatible with both fixed-angle and swing bucket centrifuge rotors.
Brand:  Thermo Scientific™ 362694_OLDNS1
Product Code. 10512802
  • 365.26 € / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes can be opened and closed with one hand due to a proprietary hinged-cap system. The tubes feature an ergonomic design, and are compatible with both fixed-angle and swing bucket centrifuge rotors.
Features:
  • Integrated leakproof cap prevents cross-contamination and lost closures
  • Clarified polypropylene combines chemical resistance and high-speed tolerances
  • Writing area and flat caps for sample identification
  • Volume markings and graduations
  • DNase/RNase free
  • Product raw materials are biocompatible per USP Class VI test
  • Product is non-cytotoxic and non-pyrogenic
  • Available in sterilized (SAL 10-6)
  • Max RCF: 8500xg for the fixed-angle rotor, 7000xg for swing bucket rotor***
  • Operating Temperature: -20 C to 37C
  • Shelf life: 2 years
***When fully supported by conical rotor cavity or conical adaptor
Ordering Information:
Tubes are supplied in a bulk pack (sleeve) or a rack.