Learn More
Thermo Scientific™ Nunc™ FluoroNunc™/LumiNunc™ 96-Well Plates

Use these well plates for quantitative and qualitative solid phase immunoassays (e.g., FLISA) and binding assays.
186.84 € - 296.00 €
Specifications
Binding Property | Hydrophilic |
---|---|
Format | Standard |
Lid | With Lid |
Culture Area | 2.5 cm2 |
Array | 8 x 12 |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
10366281
![]() |
Thermo Scientific™
437796 |
White |
296.00 €
Pack of 60 |
In Stock
Log in to see stock. ![]() |
||||||
10634412
|
Thermo Scientific™
437958 |
Clear | N/A | N/A | N/A | |||||
Description
Use Thermo Scientific™ Nunc FluoroNunc/LumiNunc 96-Well Solid plates for quantitative and qualitative solid phase immunoassays (e.g., FLISA) and binding assays. Thermo Scientific Nunc Immuno plates offer lot-to-lot reliability, well-to-well reproducibility, and are backed by industry-leading quality standards and certifications.
- Plate has SBS footprint—fits standard equipment
- Available with flat-bottom or C-shaped wells
- Choice of PolySorp™ or MaxiSorp™ surface
- Offered as black, white or clear plates
- Certified binding homogeneity/reproducibility
Choice of Surface
PolySorp - Hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains
MaxiSorp - Hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character. Available with or without lot specific
- performance testing via an IgG binding assay (Well to Well %CV less than 5% for clear plates and less than 10% for colored plates). Tested items come with a certificate
White
- Gives maximum reflection
- Minimum autofluorescence and autoluminescence
Black
- Minimizes background in fluorescence reading
- Minimizes back-scattered light
- Minimizes crosstalk
Clear
- For time-resolved fluorescence
Compatible with:
Compatible with lids for MicroWell™ plates, sealing tape and breathable membrane
Specifications
Hydrophilic | |
With Lid | |
8 x 12 | |
Nunc | |
Polystyrene | |
250 μL | |
60 | |
None |
Standard | |
2.5 cm2 | |
Adsorption | |
Microplate | |
96 | |
MaxiSorp | |
5 | |
Antibodies (Glycoproteins) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.