Learn More
Thermo Scientific™ Nunc™ Immuno TSP Lids
Simplify hybridoma screening with Thermo Scientific™ Nunc™ Immuno TSP lids for antibody production. Use for bacterial inoculations of 96-well plates.
Brand: Thermo Scientific™ 445497
Description
Simplify hybridoma screening with Thermo Scientific™ Nunc™ Immuno TSP lids for antibody production. Use in conjunction with the Thermo Scientific™ Nunc™ OmniTray™ for bacterial inoculations of 96-well plates. Nunc Immuno TSP Lids feature uniform adsorption properties for efficient screening.
- Ideal for screening of hybridoma cultures for antibody production
- For hybridoma screening mounted in the OmniTray
- Uniform adsorption properties
- 96-pin lid for bacterial inoculations of 96-well plates
Specifications
Clear | |
Polystyrene | |
127.4 mm | |
1 | |
85.3 mm | |
Plate Lid | |
96-well Plate, OmniTray |
Inoculation Lid | |
Sterile | |
50 | |
Case of 50 | |
Hybridoma screening | |
Nunc |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.