missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide System

Grow anchorage dependent cells right on the microscope slide using the unique removable chamber of the Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide System.

Brand:  Thermo Scientific™ 178599

 View more versions of this product

Product Code. 10507401

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Grow, stain, and examine cells directly on the Thermo Scientific™ Nunc™ Lab-Tek™ Chamber Slide™, which consists of a polystyrene media chamber attached to a microscopic slide. Slide surface is specially treated for adherent cell culture. Choose either specialty washed Soda Lime Glass slide or cell attachment optimized Permanox Plastic slide. Media chamber is easily removable without the need for a tool.

  • Broad range of formats and well number to meet application needs
  • Available with a Permanox solvent-resistant plastic slide for adherent cell culture
  • Writing surface provides easy identification of samples
  • Save time and reagents by growing cells directly on microscope slide
  • In 1, 2, 4, 8 and 16 well configurations; all measure 25 × 75mm
  • Medical-grade silicone sealing gasket can be removed for cover slipping
  • No cell transfer needed prior to visualization/staining
  • Useful for viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology
  • Suitable for use with fluorescent labels
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications

Specifications

16
Polystyrene
Cell Culture
0.1 to 0.2 mL
16
Sterile
0.4 cm2
16-well Chamber Slide w/ removable wells
Soda Lime Glass
Fitted clear polystyrene lid without handle
75 x 25 mm
Clear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.