missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Lab-Tek™ II CC2™ Chamber Slide System

Product Code. 13093043
Click to view available options
Wells:
1
2
4
8
Material:
CC2 Glass (mimics polylysine)
Description:
1-well Chamber Slide w/ removable well
2-well Chamber Slide w/ removable wells
4-well Chamber Slide w/ removable wells
8-well Chamber Slide w/ removable wells
Unit Size:
Pack of 16
Pack of 96
This item is not returnable. View return policy

Product Code. 13093043

Brand: Thermo Scientific™ 154941

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enable cell growth directly on a glass microscope slide with the Thermo Scientific™ Nunc™ Lab-Tek™ II CC2™ Chamber Slide System.

Enable cell growth (e.g. neurons) directly on an aminated glass slide with the Thermo Scientific™ Nunc™ Lab-Tek™ II CC2™ Chamber Slide System, which features a chemically modified CC2 growth surface that mimics the characters of polylysine coating. Available in 1-, 2-, 4-, and 8-well formats with removable media chambers.

  • Remains stable and consistent for over one year without refrigeration
  • Tabbed cover allows gas exchange while minimizing evaporation
  • Sterile
  • Removal tool separates chamber from glass
  • Provides binding sites optimal for fastidious cells
  • Light blue frosted writing area

Includes: Removal tool

TRUSTED_SUSTAINABILITY

Specifications

Wells 8
Culture Area 0.7 cm2
Lid Material Polystyrene
Description 8-well Chamber Slide w/ removable wells
Surface Treatment Cell Culture
Material CC2 Glass (mimics polylysine)
Volume (Metric) Working 0.2 to 0.5 mL
Includes Clear polystyrene lid with handles on the top and side for aseptic manipulations
No. per Pack 16
Dimensions (LxW) 74 x 25 mm
Sterility Sterile
Closure Color Clear
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.