missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Latch Rack for 1.8 mL Externally-Threaded Universal Tubes
GREENER_CHOICE

Versatile tubes for use inside a large storage system, or in smaller scale for traditional cryostorage in dewars.

180.00 € - 295.00 €

Specifications

Capacity (Metric) 1.8 mL (Tube)
Color Beige
Type Latch Rack
For Use With (Equipment) Cryobank
Holds Externally-Threaded Universal Tubes
View More Specs

Products 2
Product Code Brand Barcode Price Quantity & Availability  
Product Code Brand Barcode Price Quantity & Availability  
13426123
GREENER_CHOICE
View Documents
Thermo Scientific™
331830
None
180.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
 
15553694
GREENER_CHOICE
View Documents
Thermo Scientific™
331830-BR
2D, linear and human readable
295.00 €
Pack of 10
In Stock
to see stock.
Add to basket
 
Description

Description

Use versatile Thermo Scientific™ Nunc™ 1.8 mL Externally-threaded Universal Tubes inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars. Available racked or in bulk, with 2D barcodes or non-coded.

Scalable

  • Tubes and rack are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cryostorage boxes made for 10x10 arrays
  • External threads limit contamination
  • Automation-pickable in Universal Latch Rack

Innovative Universal Cap Design

  • Single-piece construction with molded-in gasket
  • Cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen

Secure Tracking

  • Available with a permanently-bonded, unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
  • 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
  • Scan the tube to access sample information and identify the contents
  • 374502NOV is featured with 1D (linear) barcode and human readable on the side that matches 2D barcode on the bottom of the tube

Stringent Quality Control

  • Every 2D barcoded storage tube is scanned to ensure readability
  • Barcodes are checked against database of previously-assigned codes to prevent duplicates
  • Every tube has a Sterility Assurance Level of 106 and is made with class VI medical grade resins
Specifications

Specifications

1.8 mL (Tube)
Latch Rack
Externally-Threaded Universal Tubes
1/Pk., 10/Cs.
Beige
Cryobank
10 empty racks
Non-sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.