missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 96 Well Plate, Non-Treated, Surface, No Lid, Non-sterile, Pack of 10
Clear polystyrene plates are ideal for optical measurements. Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment.
Brand: Thermo Scientific™ 260836
231.14 EUR valid until 2024-11-30
Use promo code "21681" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Specifications
Custom Barcoding Available | |
Without Lid | |
8 x 12 | |
Nunc | |
Microplate | |
400 μL | |
Non-sterile | |
Flat Bottom (F-well) | |
10 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
96F well plate, Non-Treated, clear, without lid, Non-sterile (Pack of 10, Case of 180) | |
10 Pk., 180 Case | |
Clear | |
Polystyrene | |
96 | |
Non-treated | |
180 |