Learn More
Thermo Scientific™ 96 Well Plate, Non-Treated Surface, Pack of 25
These microplates are optimized for robotics, automated readers and liquid handling systems. Ideal for antibiotic screens, cell-based assays and screening compounds.
Brand: Thermo Scientific™ 266120
233.15 EUR valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : NEW
Additional Details : Weight : 6.26400kg
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Specifications
Custom Barcoding Available | |
With Lid | |
8 x 12 | |
Nunc | |
Microplate | |
96 | |
Non-treated | |
100 | |
100/Cs. |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
96F well plate, Non-Treated, clear, with lid, Sterile (Pack of 25, Case of 100) | |
25/Pk., 100/Cs. | |
Clear | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
25 |