Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates

Obtain consistent results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates, optimized for robotics, automated readers and liquid handling systems.
Brand: Thermo Scientific™ 260860
390.01 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Obtain consistent and reliable results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available

Specifications
No Barcode | |
Standard | |
96F well plate, Non-Treated, clear, with lid, Sterile (Pack of 10, Case of 160) | |
160 Cases | |
Clear | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
10 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
With Lid | |
8 x 12 | |
Nunc | |
Microplate | |
96 | |
Non-treated | |
160 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.