missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates
BIOPHARMA

Obtain consistent results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates, optimized for robotics, automated readers and liquid handling systems.

Brand:  Thermo Scientific™ 243656

197.34 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 10774957

  • 296.00 € / Pack of 108
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Obtain consistent and reliable results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.

  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Barcode
Standard
96F well plate, Non-Treated, clear, with lid, Sterile (Pack of 6, Case of 108)
108 Cases
Clear
Polystyrene
Sterile
Flat Bottom (F-well)
6
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
With Lid
8 x 12
Nunc
Microplate
96
Non-treated
108
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.