Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate Product Code.: 10072151
In Stock
to see stock.
Add to basket
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
BIOPHARMA
Product Code. 10072151
96, 1 Microplate, 50 Microplates
Click to view available options
No. of Wells:
96
No. per Pack:
1 Microplate
10 Microplates
No. per Case:
160 Microplates
50 Microplates
Product Code. 10072151
Brand: Thermo Scientific™ 136101
In Stock
to see stock.
Edge
Add to basket
Add to basket
Color: White; Sterility: Sterile; Lid: With Lid
Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ White Polystyrene Plate. White plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics.
  • Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
  • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Working volume range: 50 to 250μL/well
  • Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
  • Customized bar coding is available
TRUSTED_SUSTAINABILITY
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Certified Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
Lid With Lid
Array 8 x 12
Packaging 1/Pk., 50/Cs.
Product Line Nunc
Color White
Product Type Microplate
Skirt Style Low Flange Pinchbar
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 50 to 200 μL
Sterility Sterile
Surface Treatment Nunclon Delta
Well Shape Flat Bottom (F-well)
No. per Case 50 Microplates
No. per Pack 1 Microplate
Quantity 50/Cs.
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number185244 Certificate TypeCertificate of Compliance Date08/03/2025 Catalog Number136101
Lot Number184073 Certificate TypeCertificate of Compliance Date24/12/2024 Catalog Number136101
Lot Number178972 Certificate TypeCertificate of Compliance Date02/12/2024 Catalog Number136101
Lot Number185244 Certificate TypeCertificate of Irradiation Date28/02/2025 Catalog Number136101
Lot Number184073 Certificate TypeCertificate of Irradiation Date13/12/2024 Catalog Number136101
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate >