missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate

Click to view available options
Quantity:
160/Cs.
50/Cs.
Description
Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ Black and White Polystyrene Plate. Black and white plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics. Available with or without cell culture treatment.
- Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
- Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
- White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
- Working volume range: 50 to 250μL/well
- Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
- Customized bar coding is available

Spécification
Spécification
Barcode | Custom Barcoding Available |
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Certified | Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility |
Lid | With Lid |
Array | 8 x 12 |
Packaging | 1/Pk., 50/Cs. |
Product Line | Nunc |
Color | Black |
Product Type | Microplate |
Material | Polystyrene |
Afficher plus |
Documents
Scientific Resources
Application Notes
Application Note: Activity of Adsorbed Antibodies
File size: 171.1 kB
Activity of Adsorbed Antibodies
File size: 1.5 MB
Application Note: Colorimetric Determination of Amino Groups of Nunc CovaLink NH MicroWell Plates
File size: 238.2 kB
Technical Bulletin: Nunc FluoroNunc Modules and Plates - The principle behind Q.C. procedure
File size: 159.4 kB
Technical Bulletin: Effects of Enlarged Surface/Volume/Ratio in Solid Phase Assays Documented on the basis of Nunc StarWell Modules
File size: 220.6 kB
Technical Bulletin: Principles in Adsorption to Polystyrene
File size: 225.2 kB
Technical Bulletin: Covalent Binding of DNA to CovaLink NH Methods and Applications
File size: 113.2 kB
Technical Bulletin: Blocking Agent and Detergent in ELISA
File size: 158 kB
Product Information
Product Bulletins
Covalent Binding of DNA to CovaLink NH Methods and Applications
File size: 113.2 kB
Detergent in Polystyrene ELISA
File size: 939 kB
Comparison of Affinity-Isolated and Non-Isolated Antibodies Used as capture Antibodies in ELISA
File size: 1.4 MB
Aspects of Nunc MicroWell Certification/Negative Edge Effect in Nunc MicroWell ELISA
File size: 919 kB
Effects of Enlarged Surface/Volume/Ratio in Solid Phase Assays Documented on the basis of Nunc StarWell Modules
File size: 434.8 kB
Edge effect in Nunc MicroWell ELISA
File size: 452 kB
Principles in Adsorption to Polystyrene
File size: 5.8 MB
Blocking Agent and Detergent in ELISA
File size: 1.6 MB
Warranties
Manuals
Technical Data Sheet: Nunc F96 Microwell Plate, PS
Version: August 2015
File size: 136.1 kB
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate >
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu