Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate

Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available

Specifications
Specifications
Barcode | Custom Barcoding Available |
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Lid | Without Lid |
Description | 96F well plate, Nunclon Delta, clear, without lid, Sterile (Pack of 10, Case of 180) |
Array | 8 x 12 |
Packaging | 10/Pk., 180/Cs. |
Product Line | Nunc |
Color | Clear |
Product Type | Microplate |
Skirt Style | High Flange |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
185182 | Certificate of Irradiation | 30/04/2025 | 156545 |
185182 | Certificate of Compliance | 26/02/2025 | 156545 |
182469 | Certificate of Compliance | 03/12/2024 | 156545 |
171194 | Certificate of Compliance | 02/12/2024 | 156545 |
180759 | Certificate of Compliance | 02/12/2024 | 156545 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.