Learn More
Thermo Scientific™ Nunc™ Multidishes with Nunclon™ Supra Surface
Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.
Brand:  Thermo Scientific™ 140680
233.80 EUR valid until 2025-12-31
Use promo code "24071" to get your promotional price.
 View more versions of this product
Product Code. 17907472
  • 334.00 € / Pack of 75
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.
Features of Nunc Multidishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years
TRUSTED_SUSTAINABILITY
Specifications
Specifications
With Lid
Clear
Polystyrene
Sterile
Flat Bottom (F-well)
1 Dish
Nunc
Single-use
6
Nunclon Supra
75 Dishes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ Multidishes with Nunclon™ Supra Surface > 6 Wells
For Laboratory Use