Learn More
Thermo Scientific™ Nunc™ Rectangular Dishes
Perform molecular biology applications with non-treated Thermo Scientific™ Nunc™ Rectangular Dishes, which have ANSI™ standard dimensions and are optimized for robotics.
Brand: Thermo Scientific™ 267062
696.70 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Non-treated Thermo Scientific™ Nunc™ Rectangular Dishes with ANSI™ standard dimensions are optimized for molecular biology applications using robotic instruments.
Rectangular Dishes 1, 4 and 8-Well
- 1-well dish is ideal for HTS Petri dish applications
- 4- and 8-well dishes are ideal for assay compartmentalization, probe multiple gene sub-set
- 4-well dish holds four glass slides for storage
1-Well Format
- Convenient plate format for cell culture
- HTS Petri dish applications
4- and 8-Well Formats
- Assay compartmentalization
- Probe multiple gene subsets
- Wash Western blot strips
- 4-well dish holds four glass slides
- Useful in all areas of cell culture, including scale-up and cloning
Ordering Information:
Customized bar coding is available

Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
8 | |
Non-treated | |
Rectangular | |
100 | |
100 mm | |
59 mm |
Cell Culture Dish | |
Sterile | |
11 mL | |
Lid | |
10 | |
86 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.