missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Storage Vials with Caps
GREENER_CHOICE

Assure safe centrifugation and storage of PCR reagents, restriction enzymes, and other diagnostic and biochemical reagents or samples by using these storage vials with caps.

460.00 € - 466.00 €

Specifications

Material PPCO
Sterility Sterile
No. per Case 500
No. per Pack 500
Product Line Nunc
View More Specs

Products 2
Product Code Brand Volume (Metric) Working Price Quantity & Availability  
Product Code Brand Volume (Metric) Working Price Quantity & Availability  
10694022
GREENER_CHOICE
View Documents
Thermo Scientific™
264262
0.5 mL
460.00 €
Pack of 500
In Stock
to see stock.
Add to basket
 
10749081
GREENER_CHOICE
View Documents
Thermo Scientific™
264300
2.0 mL
466.00 €
Pack of 500
In Stock
to see stock.
Add to basket
 
Description

Description

Assure safe centrifugation and storage of PCR reagents, restriction enzymes and other biochemical reagents and samples by using Thermo Scientific™ Nunc™ Storage Vials with Caps. Available in 0.5mL and 2.0mL sizes with frosted writing area, the vials have excellent chemical resistance with leakproof closure.

Features:

  • Made from high-purity polypropylene copolymer with excellent chemical resistance
  • Leakproof high-density polyethylene screw closure
  • Centrifuged to a maximum of 13,000 x g
  • Temperature range: -40°C to 95°C
  • RNase/DNase-free

Ordering Information:

Racks 264263 and 270972 are compatible with these vials. Color coders available: 254416, 264335, 264408, 264440, 264459, 264467, 264491.

Specifications

Specifications

PPCO
500
Nunc
Screw Cap
Certificate of Analysis (COA), Certificate of Conformance (COC)
Sterile
500
Polyethylene
500 Cs.
Storage Vial with Cap
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.