missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 384 Well White Plate, Optically Clear Polymer Bottom, Pack of 10

Color:White, Clear Bottom; Sterility:Sterile; Lid:With Lid; Type: Optical Polymer Base

Brand:  Thermo Scientific™ 142762

669.00 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10060601

  • 892.00 € / Pack of 30
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Nunc 384 Well Optical Bottom Plates are ideal for microscopic applications. The white microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence.

Rounded-square wells provide maximum access for dispensers and readers.
  • Poly- D-Lysine and Collagen coating is ideal for cells with low adherence or growth—the uniform coating creates a positive charge on the surface that ensures cell attachment, growth and differentiation.
  • Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment and are ideal for adding coating in your own lab
  • Polystyrene upper structure with either a #1.5 cover glass or polystyrene base
  • Working volume: 10 to 120μL
  • Customized bar coding is available
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Custom Barcoding Available
With Lid
10 Pk., 30 Cases
White
Polystyrene
384
Sterile
Flat Bottom (F-well)
10
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
16 x 24
Nunc
Optical Polymer Base
120 μL
50 to 200 μL
Cell Culture
30
Case of 30
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Schematics & Diagrams

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1425422 Certificate TypeCertificate of Compliance Date08/04/2025 Catalog Number142762
Lot Number1421423 Certificate TypeCertificate of Compliance Date28/01/2025 Catalog Number142762
Lot Number1421423 Certificate TypeCertificate of Processing Date28/03/2025 Catalog Number142762
Lot Number1416215 Certificate TypeCertificate of Processing Date12/02/2025 Catalog Number142762
Lot Number1276495 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number142762
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ 384 Well White Plate, Optically Clear Polymer Bottom, Pack of 10 > 384 Well Optical Bottom Plates, Polystyrene Polymer Base, Cell Culture Treated, White, with lid, Sterile

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.