missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Tris-Acetate Welcome Pack, 3-8%

The NuPAGE Tris-Acetate Welcome Pack provides all of the necessary NuPAGE Tris-Acetate gels, buffers, and reagents needed to begin using the Mini Gel Tank.

Brand:  Invitrogen™ EA0375B

851.34 EUR valid until 2024-12-31
Use promo code "21959" to get your promotional price.



Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15864042

  • 914.00 € / Each
Estimated Shipment: 12-08-2024
to see stock.
Add to basket

Description

Description

The NuPAGE Tris-Acetate Welcome Pack provides all of the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading, and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.

The NuPAGE Tris-Acetate Welcome Pack contains:
• Mini Gel Tank (A25977)
• NuPAGE Tris-Acetate mini gels (2 boxes, 20 gels)
• NuPAGE Tris-Acetate SDS Running Buffer, 20X (LA0041)
• NuPAGE LDS Sample Buffer, 4X (NP0007)
• NuPAGE Sample Reducing Agent, 10X (NP0009)
• HiMark Pre-stained Protein Standard (LC5699)

About the Mini Gel Tank
The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.

About NuPAGE Tris-Acetate gels
NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. NuPAGE Tris-Acetate gels are high performance polyacrylamide gels that simulate the denaturing or the native conditions of the traditional laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins compared to traditional Tris-glycine SDS-PAGE gels.

For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer buffer (NP0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Services

Up to 12 months

Specifications

Specifications

Mini Gel Tank
Mini
Tris-Acetate
12-well
1 Welcome Pk.
3 to 8%
1.0 mm
40 to 500 kDa
NuPAGE™
Approved for shipment at room temperature and wet ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers