Learn More
VICI Valco Instruments™ Valco Stainless Nuts
These nuts are for use with all standard Valco internal fittings and most valves
Brand:  VICI Valco Instruments™ JR-ZN1-5
Product Code. 12903531
  • 47.35 € / Pack of 5
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Excellent fit to Valco fitting details
  • Different lengths available
  • They may be used with fittings from other manufacturers as well.
Specifications
Specifications
1/16 in.
10 to 32
20,000 psi
5
11 mm