Learn More
Thermo Scientific™ Nutrient Broth (Bottled)
Cultivate non-fastidious microorganisms for testing food, milk and dairy products using these nutritious, ready prepared bottles.
Brand: Thermo Scientific™ BO0210B
Description
Cultivate non-fastidious microorganisms for testing food, milk and dairy products with Thermo Scientific™ Oxoid™ Nutrient Broth ready prepared bottles. This non-selective, fluid medium contains beef extracts combined with peptone, yeast extracts and sodium chloride to provide the nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.
Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared bottles of Nutrient Broth.
- Suitable for general cultivation of less fasidious microorganisms.
- Convenient: the pre-filled bottles are ready for immediate use.
- Simple: easy-to-use with minimum training.
- Long shelf life: up to 12 months.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Straw, clear | |
Nutrient Broth (Bottled) | |
United Kingdom | |
3.0g + 0.3g | |
24/Pk. |
2 to 25°C | |
Bijou Bottles | |
Microbiology Media | |
Twenty four bijou bottles with plastic screw cap closures in a box |